Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011104.3 | NP_035234.1 | 10 - 11 | 2248 | 81 | |
GenBank mRNA | AF028009.1 | - | 10 - 11 | 1449 | 81 | |
AF325507.1 | - | 10 - 11 | 1440 | 81 | ||
AK082438.1 | - | 10 - 11 | 2249 | 81 | ||
AK085351.1 | - | 11 - 12 | 1660 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mus musculus Probe spans exons |