Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019924.2 | NP_064308.2 | 3 - 4 | 423 | 73 | |
GenBank mRNA | AF074714.1 | - | 3 - 4 | 395 | 73 | |
AK089467.1 | - | 3 - 4 | 414 | 73 | ||
AK154674.1 | - | 3 - 4 | 422 | 73 | ||
BC012964.1 | - | 3 - 4 | 433 | 4160873 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |