Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038589.2 | NP_001033678.1 | 13 - 14 | 1335 | 127 | |
NM_021522.4 | NP_067497.2 | 14 - 15 | 1440 | 127 | ||
GenBank mRNA | AB034633.1 | - | 14 - 15 | 1219 | 127 | |
AK029977.1 | - | 14 - 15 | 1369 | 127 | ||
AK045909.1 | - | 14 - 15 | 1360 | 127 | ||
AK170933.1 | - | 1434 | 127 | |||
AK172477.1 | - | 14 - 15 | 1426 | 127 | ||
BC005571.1 | - | 14 - 15 | 1359 | 3157723 | 127 | |
BC050197.1 | - | 13 - 14 | 1253 | 30006882 | 127 | |
CN531861.1 | - | 2 - 3 | 114 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |