Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038589.2 | NP_001033678.1 | 5 - 6 | 583 | 100 | |
NM_021522.4 | NP_067497.2 | 6 - 7 | 688 | 100 | ||
GenBank mRNA | AB034633.1 | - | 6 - 7 | 467 | 100 | |
AK011322.1 | - | 6 - 7 | 624 | 100 | ||
AK029977.1 | - | 6 - 7 | 617 | 100 | ||
AK045909.1 | - | 6 - 7 | 608 | 100 | ||
AK160215.1 | - | 6 - 7 | 622 | 100 | ||
AK170933.1 | - | 682 | 100 | |||
AK172477.1 | - | 6 - 7 | 675 | 100 | ||
BC005571.1 | - | 6 - 7 | 607 | 3157723 | 100 | |
BC050197.1 | - | 5 - 6 | 501 | 30006882 | 100 | |
CB520876.1 | - | 5 - 6 | 490 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |