Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021485.2 | NP_067460.1 | 12 - 13 | 1041 | 112 | |
GenBank mRNA | AJ007938.1 | - | 12 - 13 | 1041 | 112 | |
AK014412.1 | - | 12 - 13 | 1120 | 112 | ||
AK090093.1 | - | 1096 | 112 | |||
AK156361.1 | - | 12 - 13 | 1095 | 112 | ||
BC005694.1 | - | 4 - 5 | 372 | 112 | ||
BC051631.1 | - | 12 - 13 | 1347 | 112 | ||
BC066842.1 | - | 11 - 12 | 1458 | 30468251 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |