Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178650.3 | NP_848765.2 | 7 - 8 | 700 | 76 | |
GenBank mRNA | AK040487.1 | - | 7 - 8 | 701 | 76 | |
AK075803.1 | - | 7 - 8 | 687 | 76 | ||
AK146426.1 | - | 7 - 8 | 694 | 76 | ||
AK153836.1 | - | 7 - 8 | 698 | 76 | ||
BC094901.1 | - | 6 - 7 | 675 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |