Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081080.1 | NP_001074549.1 | 9 - 10 | 3195 | 74 | |
GenBank mRNA | AK122230.1 | - | 5 - 6 | 646 | 74 | |
BC099524.1 | - | 3 - 4 | 278 | 74 | ||
BC137913.1 | - | 9 - 10 | 3201 | 8860932 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |