Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081080.1 | NP_001074549.1 | 1 - 2 | 269 | 145 | |
GenBank mRNA | BC011226.1 | - | 1 - 2 | 264 | 145 | |
BC032205.1 | - | 1 - 2 | 283 | 145 | ||
BC137913.1 | - | 1 - 2 | 275 | 8860932 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |