Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081080.1 | NP_001074549.1 | 3 - 4 | 2144 | 93 | |
GenBank mRNA | BC011226.1 | - | 3 - 4 | 2139 | 93 | |
BC019710.1 | - | 1 - 2 | 625 | 94 | ||
BC032205.1 | - | 3 - 4 | 2158 | 93 | ||
BC038053.1 | - | 1 - 2 | 553 | 93 | ||
BC137913.1 | - | 3 - 4 | 2150 | 8860932 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |