Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008000.2 | NP_032026.2 | 3 - 4 | 467 | 128 | |
GenBank mRNA | M32054.1 | - | 3 - 4 | 467 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |