Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Safb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029979.2 | NP_001025150.2 | 18 - 19 | 2788 | 80 | |
GenBank mRNA | AK031971.1 | - | 1 - 2 | 65 | 80 | |
BC028827.1 | - | 12 - 13 | 1502 | 80 | ||
BC050855.1 | - | 18 - 19 | 2722 | 80 | ||
BC057039.1 | - | 16 - 17 | 1918 | 80 | ||
BC107016.2 | - | 1 - 2 | 73 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |