Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175488.6 | NP_780697.2 | 5 - 6 | 556 | 153 | |
GenBank mRNA | AK029803.1 | - | 5 - 6 | 557 | 153 | |
AK030198.1 | - | 5 - 6 | 545 | 153 | ||
AK030205.1 | - | 2 - 3 | 206 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |