Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033289.2 | NP_001028461.2 | 8 - 9 | 1915 | 64 | |
GenBank mRNA | AF139193.1 | - | 3 - 4 | 268 | 64 | |
AK077026.1 | - | 8 - 9 | 2007 | 64 | ||
AK138779.1 | - | 8 - 9 | 2292 | 64 | ||
BC103685.1 | - | 8 - 9 | 1915 | 40048593 | 64 | |
BC104737.1 | - | 8 - 9 | 1915 | 40048591 | 64 | |
BC105676.1 | - | 8 - 9 | 1914 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |