Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081287.2 | NP_001074756.2 | 10 - 11 | 1128 | 97 | |
NM_001161620.1 | NP_001155092.1 | 10 - 11 | 1128 | 97 | ||
GenBank mRNA | AK012883.1 | - | 10 - 11 | 1103 | 97 | |
AK036956.1 | - | 10 - 11 | 1099 | 97 | ||
AK078849.1 | - | 10 - 11 | 1127 | 97 | ||
BC117550.1 | - | 9 - 10 | 975 | 40105182 | 97 | |
BC118058.1 | - | 9 - 10 | 975 | 40105180 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |