Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010603.6 | NP_034733.4 | 2 - 3 | 455 | 102 | |
GenBank mRNA | AK017299.1 | - | 2 - 3 | 456 | 102 | |
BC115970.1 | - | 1 - 2 | 35 | 40106507 | 102 | |
BC116677.1 | - | 1 - 2 | 35 | 40106509 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |