Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007580.1 | NP_001007581.1 | 20 - 21 | 3015 | 59 | |
GenBank mRNA | BC076625.1 | - | 20 - 21 | 3015 | 30641582 | 59 |
BC085176.1 | - | 20 - 21 | 3013 | 30626050 | 59 | |
DQ192038.1 | - | 20 - 21 | 3060 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |