Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mndal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170853.1 | NP_001164324.1 | 671 | 95 | ||
GenBank mRNA | AK087931.1 | - | 3 - 4 | 500 | 95 | |
AK136391.1 | - | 673 | 95 | |||
FJ594740.1 | - | 3 - 4 | 570 | 95 | ||
FJ594741.1 | - | 579 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus 1700 Inventoried Probe spans exons |