Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207268.3 | NP_997151.2 | 1 | 2704 | 105 | |
GenBank mRNA | AK029877.1 | - | 1 | 2656 | 105 | |
BB667164.1 | - | 1 | 210 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |