Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029402.3 | NP_083678.2 | 1 - 2 | 171 | 111 | |
GenBank mRNA | AK004875.2 | - | 1 - 2 | 171 | 111 | |
AK028589.1 | - | 1 - 2 | 140 | 111 | ||
AK144693.1 | - | 1 - 2 | 144 | 111 | ||
AK160597.1 | - | 1 - 2 | 156 | 111 | ||
AK220287.1 | - | 1 - 2 | 146 | 111 | ||
BC026779.1 | - | 1 - 2 | 72 | 4018298 | 111 | |
BC027428.1 | - | 1 - 2 | 134 | 3983461 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons |