Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177409.3 | NP_803128.1 | 4 - 5 | 526 | 72 | |
GenBank mRNA | AK082751.1 | - | 4 - 5 | 527 | 72 | |
AY029530.1 | - | 4 - 5 | 463 | 72 | ||
BC018212.1 | - | 4 - 5 | 511 | 3980004 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |