Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027974.1 | NP_082250.1 | 5 - 6 | 1029 | 69 | |
GenBank mRNA | AK006489.1 | - | 5 - 6 | 968 | 69 | |
EU520262.1 | - | 5 - 6 | 921 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |