Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021287.1 | NP_067262.1 | 20 - 21 | 4729 | 67 | |
GenBank mRNA | AK147671.1 | - | 20 - 21 | 4745 | 67 | |
AK172931.1 | - | 10 - 11 | 3038 | 67 | ||
BC079860.1 | - | 20 - 21 | 4729 | 6834745 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |