Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021287.1 | NP_067262.1 | 32 - 33 | 6893 | 96 | |
GenBank mRNA | AK147671.1 | - | 32 - 33 | 6909 | 96 | |
AK172931.1 | - | 22 - 23 | 5202 | 96 | ||
BC048851.1 | - | 10 - 11 | 1711 | 96 | ||
BC079628.1 | - | 10 - 11 | 1845 | 96 | ||
BC079860.1 | - | 32 - 33 | 6893 | 6834745 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |