Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013456.2 | NP_038484.1 | 2 - 3 | 314 | 80 | |
GenBank mRNA | AF093775.1 | - | 2 - 3 | 289 | 80 | |
BC111890.1 | - | 2 - 3 | 280 | 40060598 | 80 | |
BY132390.1 | - | 2 - 3 | 314 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |