Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290818.1 | NP_001277747.1 | 5 - 6 | 649 | 62 | |
NM_130863.2 | NP_570933.1 | 6 - 7 | 721 | 62 | ||
GenBank mRNA | AF333028.1 | - | 5 - 6 | 441 | 62 | |
AK155691.1 | - | 5 - 6 | 650 | 62 | ||
BC003196.1 | - | 5 - 6 | 437 | 62 | ||
BC033272.1 | - | 5 - 6 | 506 | 62 | ||
BC053922.1 | - | 6 - 7 | 454 | 6309555 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |