Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033128.3 | NP_001028300.1 | 3 - 4 | 186 | 103 | |
GenBank mRNA | AK037753.1 | - | 3 - 4 | 172 | 103 | |
AK045850.1 | - | 3 - 4 | 183 | 103 | ||
AK161343.1 | - | 3 - 4 | 180 | 103 | ||
AK168901.1 | - | 3 - 4 | 187 | 103 | ||
BC076577.1 | - | 3 - 4 | 175 | 103 | ||
BY063920.1 | - | 3 - 4 | 187 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |