Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033128.3 | NP_001028300.1 | 6 - 7 | 539 | 92 | |
GenBank mRNA | AK037753.1 | - | 6 - 7 | 525 | 92 | |
AK045850.1 | - | 6 - 7 | 537 | 92 | ||
AK161343.1 | - | 6 - 7 | 533 | 92 | ||
AK168901.1 | - | 6 - 7 | 540 | 92 | ||
BC076577.1 | - | 6 - 7 | 528 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |