Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018775.4 | NP_061245.3 | 8 - 9 | 1582 | 83 | |
GenBank mRNA | AK138630.1 | - | 8 - 9 | 1580 | 83 | |
AK141520.1 | - | 8 - 9 | 1571 | 83 | ||
AK141816.1 | - | 8 - 9 | 1536 | 83 | ||
AK162371.1 | - | 2 - 3 | 357 | 83 | ||
BC005421.1 | - | 5 - 6 | 917 | 83 | ||
BC065081.1 | - | 8 - 9 | 1516 | 6832495 | 83 | |
U88873.1 | - | 4 - 5 | 802 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |