Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009044.2 | NP_033070.2 | 3 - 4 | 588 | 147 | |
GenBank mRNA | AK031627.1 | - | 3 - 4 | 387 | 147 | |
BC139447.1 | - | 3 - 4 | 528 | 8862469 | 147 | |
BC139448.1 | - | 3 - 4 | 528 | 8862470 | 147 | |
BC139770.1 | - | 3 - 4 | 530 | 40111461 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |