Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009044.2 | NP_033070.2 | 8 - 9 | 1201 | 128 | |
GenBank mRNA | AK031627.1 | - | 8 - 9 | 999 | 128 | |
BC139447.1 | - | 8 - 9 | 1141 | 8862469 | 128 | |
BC139448.1 | - | 8 - 9 | 1141 | 8862470 | 128 | |
BC139770.1 | - | 8 - 9 | 1143 | 40111461 | 128 | |
X15842.1 | - | 8 - 9 | 1323 | 128 | ||
X60271.1 | - | 8 - 9 | 1200 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |