Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025572.1 | NP_001020743.1 | 1 - 2 | 158 | 84 | |
GenBank mRNA | AK012645.1 | - | 1 - 2 | 151 | 84 | |
AK013457.1 | - | 1 - 2 | 202 | 84 | ||
AK133715.1 | - | 1 - 2 | 157 | 84 | ||
AK153850.1 | - | 1 - 2 | 136 | 84 | ||
BC050185.1 | - | 1 - 2 | 138 | 84 | ||
BC064777.1 | - | 1 - 2 | 142 | 84 | ||
BC080825.1 | - | 1 - 2 | 150 | 84 | ||
BC094292.1 | - | 1 - 2 | 148 | 84 | ||
CF907970.1 | - | 1 - 2 | 136 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |