Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252192.1 | NP_001239121.1 | 9 - 10 | 1443 | 106 | |
NM_010164.2 | NP_034294.2 | 10 - 11 | 1559 | 106 | ||
GenBank mRNA | AK042412.1 | - | 9 - 10 | 1366 | 106 | |
AK153788.1 | - | 10 - 11 | 1565 | 106 | ||
AK162111.1 | - | 2 - 3 | 300 | 106 | ||
BC060260.1 | - | 10 - 11 | 1559 | 6848408 | 106 | |
BC066860.1 | - | 10 - 11 | 1213 | 30089692 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |