Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213733.2 | NP_998898.1 | 1 - 2 | 225 | 100 | |
GenBank mRNA | AK144651.1 | - | 1 - 2 | 198 | 100 | |
AK150313.1 | - | 1 - 2 | 226 | 100 | ||
AK156136.1 | - | 1 - 2 | 226 | 100 | ||
AK170509.1 | - | 1 - 2 | 226 | 100 | ||
BC069933.1 | - | 1 - 2 | 220 | 4008091 | 100 | |
BC080707.1 | - | 1 - 2 | 98 | 100 | ||
BU148687.1 | - | 1 - 2 | 227 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |