Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081338.1 | NP_001074807.1 | 14 - 15 | 1713 | 98 | |
GenBank mRNA | AK049859.1 | - | 14 - 15 | 1768 | 98 | |
AK220350.1 | - | 12 - 13 | 1461 | 98 | ||
BC116639.1 | - | 13 - 14 | 1672 | 40061282 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |