Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019938.3 | NP_064322.1 | 6 - 7 | 945 | 82 | |
GenBank mRNA | AB029919.1 | - | 6 - 7 | 945 | 82 | |
AK133053.1 | - | 5 - 6 | 1001 | 82 | ||
BC138245.1 | - | 6 - 7 | 897 | 8861264 | 82 | |
BC138246.1 | - | 6 - 7 | 897 | 8861265 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |