Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205330.1 | NP_001192259.1 | 15 - 16 | 3168 | 98 | |
NM_001205332.1 | NP_001192261.1 | 14 - 15 | 3054 | 98 | ||
NM_008633.4 | NP_032659.2 | 15 - 16 | 3168 | 98 | ||
GenBank mRNA | AK004694.2 | - | 7 - 8 | 1440 | 98 | |
AK087930.1 | - | 7 - 8 | 1453 | 98 | ||
AK146790.1 | - | 7 - 8 | 2433 | 98 | ||
AK166558.1 | - | 6 - 7 | 867 | 98 | ||
BC044654.1 | - | 15 - 16 | 3078 | 5400526 | 98 | |
BC050893.1 | - | 14 - 15 | 2929 | 5706356 | 98 | |
BC055332.1 | - | 15 - 16 | 3049 | 6407218 | 98 | |
BC055364.1 | - | 14 - 15 | 3008 | 30057231 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |