Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205330.1 | NP_001192259.1 | 17 - 18 | 3367 | 74 | |
NM_001205332.1 | NP_001192261.1 | 16 - 17 | 3253 | 74 | ||
GenBank mRNA | AK004694.2 | - | 9 - 10 | 1639 | 74 | |
AK087930.1 | - | 9 - 10 | 1652 | 74 | ||
AK146790.1 | - | 9 - 10 | 2632 | 74 | ||
AK166558.1 | - | 8 - 9 | 1066 | 74 | ||
BC042645.1 | - | 8 - 9 | 2533 | 5357023 | 74 | |
BC050893.1 | - | 16 - 17 | 3128 | 5706356 | 74 | |
BC055332.1 | - | 17 - 18 | 3248 | 6407218 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |