Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007896.3 | NP_031922.1 | 3 - 4 | 400 | 156 | |
GenBank mRNA | AK146206.1 | - | 3 - 4 | 318 | 156 | |
AK154245.1 | - | 3 - 4 | 344 | 156 | ||
BC043333.1 | - | 3 - 4 | 364 | 156 | ||
BC052405.1 | - | 2 - 3 | 275 | 156 | ||
BC052728.1 | - | 311 | 156 | |||
BC064444.1 | - | 311 | 6815724 | 156 | ||
U51196.1 | - | 3 - 4 | 330 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |