Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025631.3 | NP_079907.2 | 13 - 14 | 1302 | 62 | |
GenBank mRNA | AK009346.1 | - | 13 - 14 | 1274 | 62 | |
AK009504.1 | - | 13 - 14 | 1277 | 62 | ||
AK009619.1 | - | 13 - 14 | 1275 | 62 | ||
AK009754.1 | - | 9 - 10 | 878 | 62 | ||
AK009776.1 | - | 13 - 14 | 1260 | 62 | ||
AK009794.1 | - | 13 - 14 | 1262 | 62 | ||
AK009871.1 | - | 13 - 14 | 1271 | 62 | ||
AK010130.1 | - | 13 - 14 | 1257 | 62 | ||
AK010154.1 | - | 13 - 14 | 1270 | 62 | ||
AK010157.1 | - | 13 - 14 | 1301 | 62 | ||
AK010215.1 | - | 13 - 14 | 1271 | 62 | ||
AK028156.1 | - | 13 - 14 | 1273 | 62 | ||
AK028158.1 | - | 13 - 14 | 1276 | 62 | ||
BC116681.1 | - | 13 - 14 | 1257 | 40106489 | 62 | |
BC116727.1 | - | 13 - 14 | 1257 | 40106498 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |