Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285988.1 | NP_001272917.1 | 12 - 13 | 1798 | 120 | |
NM_001285989.1 | NP_001272918.1 | 12 - 13 | 1780 | 120 | ||
NM_001285990.1 | NP_001272919.1 | 11 - 12 | 1703 | 120 | ||
NM_145436.2 | NP_663411.2 | 12 - 13 | 1783 | 120 | ||
GenBank mRNA | AK148278.1 | - | 12 - 13 | 1638 | 120 | |
AK161143.1 | - | 1 - 2 | 159 | 120 | ||
AK161471.1 | - | 12 - 13 | 1649 | 120 | ||
BC157955.1 | - | 12 - 13 | 1797 | 9088128 | 120 | |
BC172100.1 | - | 12 - 13 | 1797 | 9054386 | 120 | |
BC172149.1 | - | 11 - 12 | 1702 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |