Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008916.2 | NP_032942.1 | 3 - 4 | 386 | 71 | |
GenBank mRNA | AF483522.1 | - | 3 - 4 | 306 | 71 | |
AF483523.1 | - | 3 - 4 | 306 | 71 | ||
AK052817.1 | - | 3 - 4 | 378 | 71 | ||
AK054436.1 | - | 3 - 4 | 405 | 71 | ||
AK078104.1 | - | 3 - 4 | 397 | 71 | ||
AK164290.1 | - | 3 - 4 | 375 | 71 | ||
BC014725.1 | - | 3 - 4 | 316 | 3969089 | 71 | |
BC066112.1 | - | 3 - 4 | 386 | 30361046 | 71 | |
U96724.1 | - | 3 - 4 | 417 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |