Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfatc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037177.2 | NP_001032254.1 | 3 - 4 | 1515 | 77 | |
NM_001037178.2 | NP_001032255.1 | 3 - 4 | 880 | 77 | ||
NM_001136073.2 | NP_001129545.1 | 3 - 4 | 1515 | 77 | ||
NM_001291168.1 | NP_001278097.1 | 3 - 4 | 1400 | 77 | ||
NM_001291169.1 | NP_001278098.1 | 3 - 4 | 1400 | 77 | ||
NM_001291170.1 | NP_001278099.1 | 3 - 4 | 880 | 77 | ||
NM_001291171.1 | NP_001278100.1 | 2 - 3 | 364 | 77 | ||
NM_001291172.1 | NP_001278101.1 | 3 - 4 | 1400 | 77 | ||
NM_001291173.1 | NP_001278102.1 | 2 - 3 | 364 | 77 | ||
NM_001291174.1 | NP_001278103.1 | 3 - 4 | 880 | 77 | ||
NM_001291175.1 | NP_001278104.1 | 3 - 4 | 995 | 77 | ||
NM_001291176.1 | NP_001278105.1 | 2 - 3 | 479 | 77 | ||
NM_001291177.1 | NP_001278106.1 | 3 - 4 | 995 | 77 | ||
NM_001291178.1 | NP_001278107.1 | 2 - 3 | 479 | 77 | ||
NM_001291179.1 | NP_001278108.1 | 3 - 4 | 995 | 77 | ||
NM_010899.3 | NP_035029.2 | 3 - 4 | 1515 | 77 | ||
NR_111897.1 | 2 - 3 | 364 | 77 | |||
NR_111898.1 | 2 - 3 | 479 | 77 | |||
GenBank mRNA | AF289078.1 | - | 3 - 4 | 1341 | 77 | |
AK044489.1 | - | 3 - 4 | 1401 | 77 | ||
AK081853.1 | - | 3 - 4 | 1483 | 77 | ||
AK083119.1 | - | 3 - 4 | 856 | 77 | ||
BC064006.1 | - | 3 - 4 | 1434 | 30290121 | 77 | |
EU887581.1 | - | 3 - 4 | 1281 | 77 | ||
EU887582.1 | - | 3 - 4 | 1341 | 77 | ||
EU887583.1 | - | 3 - 4 | 878 | 77 | ||
EU887584.1 | - | 3 - 4 | 978 | 77 | ||
EU887585.1 | - | 2 - 3 | 362 | 77 | ||
EU887586.1 | - | 2 - 3 | 462 | 77 | ||
EU887587.1 | - | 3 - 4 | 1281 | 77 | ||
EU887588.1 | - | 3 - 4 | 1341 | 77 | ||
EU887589.1 | - | 3 - 4 | 878 | 77 | ||
EU887590.1 | - | 3 - 4 | 978 | 77 | ||
EU887591.1 | - | 2 - 3 | 362 | 77 | ||
EU887592.1 | - | 2 - 3 | 462 | 77 | ||
EU887599.1 | - | 3 - 4 | 1281 | 77 | ||
EU887600.1 | - | 3 - 4 | 1341 | 77 | ||
EU887601.1 | - | 3 - 4 | 878 | 77 | ||
EU887602.1 | - | 3 - 4 | 978 | 77 | ||
EU887603.1 | - | 2 - 3 | 362 | 77 | ||
EU887604.1 | - | 2 - 3 | 462 | 77 | ||
U36575.1 | - | 3 - 4 | 1411 | 77 | ||
U36576.1 | - | 3 - 4 | 1411 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |