Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025827.3 | NP_080103.1 | 7 - 8 | 1383 | 73 | |
GenBank mRNA | AK004843.1 | - | 7 - 8 | 1297 | 73 | |
AK050231.1 | - | 7 - 8 | 1292 | 73 | ||
AK088348.1 | - | 6 - 7 | 1289 | 73 | ||
AK151103.1 | - | 7 - 8 | 1329 | 73 | ||
AK151406.1 | - | 5 - 6 | 897 | 73 | ||
AK152964.1 | - | 5 - 6 | 897 | 73 | ||
AK168628.1 | - | 5 - 6 | 907 | 73 | ||
AK168791.1 | - | 7 - 8 | 1329 | 73 | ||
AK168956.1 | - | 7 - 8 | 1329 | 73 | ||
AK169656.1 | - | 1297 | 73 | |||
BC049090.1 | - | 7 - 8 | 1319 | 6401542 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |