Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028071.3 | NP_082347.1 | 1 - 2 | 262 | 72 | |
GenBank mRNA | AK007994.1 | - | 1 - 2 | 224 | 72 | |
AK008085.1 | - | 1 - 2 | 224 | 72 | ||
AK010052.1 | - | 1 - 2 | 224 | 72 | ||
AK039644.1 | - | 1 - 2 | 224 | 72 | ||
AK148854.1 | - | 1 - 2 | 228 | 72 | ||
AK150888.1 | - | 1 - 2 | 224 | 72 | ||
AK151690.1 | - | 1 - 2 | 224 | 72 | ||
AK157694.1 | - | 1 - 2 | 224 | 72 | ||
AK172403.1 | - | 1 - 2 | 224 | 72 | ||
BC010249.1 | - | 1 - 2 | 217 | 3155195 | 72 | |
BC011068.1 | - | 1 - 2 | 217 | 3157387 | 72 | |
BY200873.1 | - | 1 - 2 | 266 | 72 | ||
CT010261.1 | - | 1 - 2 | 83 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |