Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026648.4 | NP_080924.1 | 7 - 8 | 1126 | 106 | |
GenBank mRNA | AK019633.1 | - | 7 - 8 | 1126 | 106 | |
BC050751.1 | - | 7 - 8 | 1045 | 6774345 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |