Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167746.1 | NP_001161218.1 | 78 - 79 | 12741 | 65 | |
GenBank mRNA | AK173343.1 | - | 35 - 36 | 5998 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |