Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291225.1 | NP_001278154.1 | 6 - 7 | 1624 | 61 | |
NM_021286.4 | NP_067261.2 | 6 - 7 | 1624 | 61 | ||
GenBank mRNA | AB206791.1 | - | 6 - 7 | 1560 | 61 | |
AK147705.1 | - | 6 - 7 | 1543 | 61 | ||
AK161576.1 | - | 1 - 2 | 34 | 61 | ||
AK165757.1 | - | 6 - 7 | 1408 | 61 | ||
AK220513.1 | - | 5 - 6 | 2254 | 61 | ||
BC053011.1 | - | 6 - 7 | 1520 | 5698186 | 61 | |
BC055345.1 | - | 6 - 7 | 1624 | 6833344 | 61 | |
D29763.1 | - | 6 - 7 | 1451 | 61 | ||
D64009.1 | - | 6 - 7 | 1424 | 61 | ||
D64010.1 | - | 6 - 7 | 1424 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |