Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013851.2 | NP_038879.2 | 8 - 9 | 1334 | 133 | |
GenBank mRNA | AF498362.1 | - | 8 - 9 | 1334 | 133 | |
AK138196.1 | - | 8 - 9 | 1335 | 133 | ||
BC141401.1 | - | 8 - 9 | 1297 | 9056014 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |