Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Git1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004144.1 | NP_001004144.1 | 10 - 11 | 1081 | 80 | |
GenBank mRNA | BC079870.1 | - | 10 - 11 | 1081 | 6831910 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |