Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100036271.mm.1 |
NM_027083.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036271.mm.1 |
NM_027083.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036271.mm.1 |
TC1100003877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036276.mm.1 |
NM_027083.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036276.mm.1 |
NM_027083.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036276.mm.1 |
TC1100003877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036274.mm.1 |
NM_027083.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036274.mm.1 |
NM_027083.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036274.mm.1 |
TC1100003877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036272.mm.1 |
NM_027083.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036272.mm.1 |
NM_027083.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036272.mm.1 |
TC1100003877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036273.mm.1 |
NM_027083.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036273.mm.1 |
NM_027083.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036273.mm.1 |
TC1100003877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036275.mm.1 |
NM_027083.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036275.mm.1 |
NM_027083.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036275.mm.1 |
TC1100003877.mm.1 |
MTA TC |